Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338921.1 | NP_001325789.1 | 5 - 6 | 1029 | 68 | |
NM_113709.5 | NP_189431.2 | 5 - 6 | 1098 | 68 | ||
GenBank mRNA | AY039585.1 | - | 5 - 6 | 977 | 68 | |
AY113073.1 | - | 5 - 6 | 949 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |