Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MES11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339038.1 | NP_001327129.1 | 3 - 4 | 966 | 124 | |
NM_113902.4 | NP_189622.1 | 3 - 4 | 1139 | 124 | ||
GenBank mRNA | BT003149.1 | - | 3 - 4 | 764 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |