Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BR6OX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113917.4 | NP_566852.1 | 5 - 6 | 1140 | 79 | |
GenBank mRNA | AB087801.1 | - | 5 - 6 | 1038 | 79 | |
AY052655.1 | - | 5 - 6 | 1090 | 79 | ||
AY063728.1 | - | 5 - 6 | 999 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |