Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 14 - 15 | 2509 | 91 | |
NM_114447.4 | NP_190164.1 | 15 - 16 | 2507 | 91 | ||
GenBank mRNA | AF030864.1 | - | 15 - 16 | 2362 | 91 | |
AF360218.1 | - | 14 - 15 | 2316 | 91 | ||
AY040062.1 | - | 14 - 15 | 2299 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |