Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 16 - 17 | 2665 | 116 | |
NM_114447.4 | NP_190164.1 | 17 - 18 | 2663 | 116 | ||
GenBank mRNA | AF030864.1 | - | 17 - 18 | 2518 | 116 | |
AF360218.1 | - | 16 - 17 | 2472 | 116 | ||
AY040062.1 | - | 16 - 17 | 2455 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |