Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 4 - 5 | 1450 | 157 | |
NM_114447.4 | NP_190164.1 | 5 - 6 | 1448 | 157 | ||
GenBank mRNA | AF030864.1 | - | 5 - 6 | 1303 | 157 | |
AF360218.1 | - | 4 - 5 | 1257 | 157 | ||
AY040062.1 | - | 4 - 5 | 1240 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |