Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339238.1 | NP_001327062.1 | 1 - 2 | 948 | 76 | |
NM_001339239.1 | NP_001327064.1 | 1 - 2 | 948 | 76 | ||
NM_114489.5 | NP_190206.2 | 1 - 2 | 948 | 76 | ||
GenBank mRNA | AY070078.1 | - | 1 - 2 | 105 | 76 | |
AY117353.1 | - | 1 - 2 | 88 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |