Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114500.5 | NP_190217.2 | 6 - 7 | 1581 | 79 | |
GenBank mRNA | FJ708732.1 | - | 6 - 7 | 1396 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |