Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114546.4 | NP_190263.1 | 1 | 2143 | 148 | |
GenBank mRNA | AK226825.1 | - | 1 | 2104 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |