Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035747.2 | NP_001030824.1 | 3 - 4 | 1117 | 109 | |
NM_001339291.1 | NP_001319701.1 | 3 - 4 | 1080 | 109 | ||
NM_001339292.1 | NP_001326351.1 | 3 - 4 | 1221 | 109 | ||
NM_001339293.1 | NP_001326353.1 | 2 - 3 | 1229 | 109 | ||
NM_001339294.1 | NP_001326354.1 | 3 - 4 | 1126 | 109 | ||
NM_001339295.1 | NP_001326352.1 | 3 - 4 | 1106 | 109 | ||
NM_001339296.1 | NP_001326350.1 | 2 - 3 | 1203 | 109 | ||
NM_114565.3 | NP_190282.2 | 3 - 4 | 1125 | 109 | ||
GenBank mRNA | BT015413.1 | - | 3 - 4 | 1052 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |