Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114573.3 | NP_566889.1 | 9 - 10 | 1780 | 111 | |
GenBank mRNA | AY099737.1 | - | 9 - 10 | 1721 | 111 | |
BT000368.1 | - | 9 - 10 | 1707 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |