Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114607.3 | NP_190323.1 | 10 - 11 | 1688 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |