Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EB1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114637.4 | NP_190353.3 | 6 - 7 | 817 | 96 | |
GenBank mRNA | AK227867.1 | - | 6 - 7 | 782 | 96 | |
BT009707.1 | - | 6 - 7 | 560 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |