Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114641.4 | NP_190357.2 | 1 - 2 | 625 | 105 | |
GenBank mRNA | BT004197.1 | - | 1 - 2 | 438 | 105 | |
BT005456.1 | - | 1 - 2 | 370 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |