Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114642.4 | NP_190358.3 | 7 - 8 | 1197 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |