Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114646.4 | NP_190362.2 | 2 - 3 | 600 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |