Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339337.1 | NP_001327142.1 | 11 - 12 | 2058 | 62 | |
NM_001339338.1 | NP_001327141.1 | 12 - 13 | 1967 | 62 | ||
NM_114647.4 | NP_190363.3 | 12 - 13 | 1967 | 62 | ||
GenBank mRNA | AY099665.1 | - | 12 - 13 | 1966 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |