Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339341.1 | NP_001325714.1 | 10 - 11 | 1457 | 99 | |
NM_114664.4 | NP_190378.2 | 10 - 11 | 1585 | 99 | ||
GenBank mRNA | AK118088.1 | - | 10 - 11 | 1549 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |