Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114679.4 | NP_190393.1 | 3 - 4 | 511 | 101 | |
GenBank mRNA | AB010916.1 | - | 3 - 4 | 435 | 101 | |
AK226519.1 | - | 3 - 4 | 373 | 101 | ||
AY072390.1 | - | 3 - 4 | 370 | 101 | ||
AY114698.1 | - | 3 - 4 | 301 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |