Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114777.4 | NP_190487.1 | 4 - 5 | 856 | 75 | |
GenBank mRNA | AK227986.1 | - | 4 - 5 | 825 | 75 | |
BT004624.1 | - | 4 - 5 | 793 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |