Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114777.4 | NP_190487.1 | 5 - 6 | 974 | 132 | |
GenBank mRNA | AK227986.1 | - | 5 - 6 | 943 | 132 | |
BT004624.1 | - | 5 - 6 | 911 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |