Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114784.3 | NP_566916.1 | 5 - 6 | 1217 | 88 | |
GenBank mRNA | AY049288.1 | - | 5 - 6 | 1165 | 88 | |
AY090286.1 | - | 5 - 6 | 1022 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |