Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339421.1 | NP_001327011.1 | 2 - 3 | 282 | 141 | |
NM_114807.5 | NP_190516.1 | 2 - 3 | 324 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |