Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125330.1 | NP_001118802.1 | 5 - 6 | 811 | 108 | |
NM_114828.6 | NP_566923.1 | 5 - 6 | 924 | 108 | ||
GenBank mRNA | AF446355.1 | - | 5 - 6 | 789 | 108 | |
AJ276124.1 | - | 5 - 6 | 791 | 108 | ||
AY097417.1 | - | 5 - 6 | 698 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |