Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339453.1 | NP_001327187.1 | 2 - 3 | 321 | 66 | |
NM_114870.4 | NP_190579.2 | 3 - 4 | 396 | 66 | ||
GenBank mRNA | BT030315.1 | - | 3 - 4 | 291 | 66 | |
EU682309.1 | - | 3 - 4 | 291 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |