Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KICP-02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339461.1 | NP_001326509.1 | 7 - 8 | 1189 | 88 | |
NM_001339462.1 | NP_001326510.1 | 6 - 7 | 1231 | 88 | ||
NM_001339463.1 | NP_001326508.1 | 7 - 8 | 1189 | 88 | ||
NM_114884.5 | NP_566931.1 | 7 - 8 | 1189 | 88 | ||
GenBank mRNA | AB061676.1 | - | 7 - 8 | 1164 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |