Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114893.3 | NP_190602.1 | 1 | 908 | 69 | |
GenBank mRNA | AK175679.1 | - | 1 | 838 | 69 | |
AK175912.1 | - | 1 | 838 | 69 | ||
BT010607.1 | - | 1 | 649 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |