Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114990.5 | NP_190699.3 | 15 - 16 | 1807 | 68 | |
GenBank mRNA | AB368490.1 | - | 15 - 16 | 1674 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |