Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114990.5 | NP_190699.3 | 16 - 17 | 1885 | 129 | |
GenBank mRNA | AB368490.1 | - | 16 - 17 | 1752 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |