Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GASA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117028.4 | NP_192698.1 | 1 - 2 | 193 | 93 | |
GenBank mRNA | AY052302.1 | - | 1 - 2 | 142 | 93 | |
U11764.1 | - | 1 - 2 | 96 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |