Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160745.1 | NP_001154217.1 | 1 - 2 | 232 | 112 | |
NM_001203765.1 | NP_001190694.1 | 1 - 2 | 312 | 112 | ||
NM_001340630.1 | NP_001319888.1 | 1 - 2 | 287 | 221 | ||
NM_001340631.1 | NP_001328831.1 | 1 - 2 | 299 | 221 | ||
NM_117040.4 | NP_192710.1 | 1 - 2 | 287 | 112 | ||
GenBank mRNA | AB071851.1 | - | 1 - 2 | 48 | 112 | |
AK229171.1 | - | 1 - 2 | 178 | 112 | ||
BT029504.1 | - | 1 - 2 | 48 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |