Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117070.5 | NP_192740.1 | 3 - 4 | 557 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |