Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSPS4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036532.3 | NP_001031609.1 | 13 - 14 | 2925 | 78 | |
NM_001340652.1 | NP_001329003.1 | 12 - 13 | 3866 | 78 | ||
NM_001340653.1 | NP_001329002.1 | 11 - 12 | 3154 | 78 | ||
NM_001340654.1 | NP_001329004.1 | 9 - 10 | 2184 | 78 | ||
NM_117080.5 | NP_192750.2 | 13 - 14 | 3775 | 78 | ||
GenBank mRNA | KJ138763.1 | - | 12 - 13 | 2747 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |