Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSPS4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036532.3 | NP_001031609.1 | 8 - 9 | 1658 | 96 | |
NM_001340652.1 | NP_001329003.1 | 7 - 8 | 2599 | 96 | ||
NM_001340653.1 | NP_001329002.1 | 6 - 7 | 1887 | 96 | ||
NM_001340654.1 | NP_001329004.1 | 4 - 5 | 917 | 96 | ||
NM_117080.5 | NP_192750.2 | 8 - 9 | 2508 | 96 | ||
GenBank mRNA | KJ138763.1 | - | 7 - 8 | 1480 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |