Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117099.6 | NP_567354.1 | 2 - 3 | 1456 | 100 | |
GenBank mRNA | AK228564.1 | - | 2 - 3 | 1326 | 100 | |
BT005751.1 | - | 2 - 3 | 1326 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Arabidopsis thaliana Probe spans exons |