Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036537.1 | NP_001031614.1 | 7 - 8 | 1047 | 61 | |
NM_117129.3 | NP_192799.1 | 7 - 8 | 1242 | 61 | ||
GenBank mRNA | AY039599.1 | - | 7 - 8 | 986 | 61 | |
AY078011.1 | - | 7 - 8 | 888 | 61 | ||
AY086286.1 | - | 7 - 8 | 964 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |