Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340684.1 | NP_001328829.1 | 3 - 4 | 884 | 67 | |
NM_117145.3 | NP_192815.1 | 4 - 5 | 845 | 67 | ||
GenBank mRNA | AY045668.1 | - | 4 - 5 | 769 | 67 | |
BT004519.1 | - | 4 - 5 | 686 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |