Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117193.3 | NP_192861.1 | 1 - 2 | 710 | 97 | |
GenBank mRNA | AY034901.1 | - | 1 - 2 | 332 | 97 | |
AY048273.1 | - | 1 - 2 | 319 | 97 | ||
AY057606.1 | - | 1 - 2 | 253 | 97 | ||
AY086294.1 | - | 1 - 2 | 469 | 97 | ||
BT002368.1 | - | 1 - 2 | 252 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |