Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117197.4 | NP_192865.1 | 4 - 5 | 676 | 78 | |
GenBank mRNA | AF370229.1 | - | 4 - 5 | 619 | 78 | |
AF428340.1 | - | 4 - 5 | 614 | 78 | ||
AF439976.1 | - | 4 - 5 | 483 | 78 | ||
AJ318019.1 | - | 4 - 5 | 522 | 78 | ||
AY085464.1 | - | 4 - 5 | 539 | 78 | ||
AY150487.1 | - | 4 - 5 | 483 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |