Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117199.2 | NP_192867.1 | 3 - 4 | 657 | 70 | |
GenBank mRNA | AF361097.1 | - | 3 - 4 | 468 | 70 | |
AF428292.1 | - | 3 - 4 | 539 | 70 | ||
BT000487.1 | - | 3 - 4 | 468 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |