Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117204.7 | NP_567378.4 | 4 - 5 | 991 | 76 | |
GenBank mRNA | AK176361.1 | - | 4 - 5 | 864 | 76 | |
GU989202.1 | - | 4 - 5 | 780 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |