Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117268.3 | NP_567384.1 | 1 - 2 | 246 | 144 | |
GenBank mRNA | AJ748742.1 | - | 1 - 2 | 230 | 144 | |
AY052271.1 | - | 1 - 2 | 239 | 144 | ||
BT001146.1 | - | 1 - 2 | 216 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |