Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341318.1 | NP_001329724.1 | 17 - 18 | 3114 | 95 | |
NM_118020.5 | NP_193637.2 | 16 - 17 | 3293 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |