Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118041.5 | NP_193656.2 | 7 - 8 | 998 | 68 | |
GenBank mRNA | AK226939.1 | - | 7 - 8 | 905 | 68 | |
AY099697.1 | - | 7 - 8 | 918 | 68 | ||
BT000298.1 | - | 6 - 7 | 794 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |