Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118912.6 | NP_194503.1 | 2 - 3 | 1340 | 63 | |
GenBank mRNA | AY142525.1 | - | 2 - 3 | 1147 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |