Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341891.1 | NP_001320078.1 | 4 - 5 | 1234 | 86 | |
NM_001341893.1 | NP_001328004.1 | 3 - 4 | 1320 | 86 | ||
NM_001341895.1 | NP_001328006.1 | 2 - 3 | 1520 | 86 | ||
NM_118935.3 | NP_567792.1 | 3 - 4 | 1434 | 86 | ||
GenBank mRNA | AF367337.1 | - | 3 - 4 | 1188 | 86 | |
BT002238.1 | - | 3 - 4 | 1178 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |