Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118949.1 | NP_194538.1 | 1 - 2 | 136 | 71 | |
GenBank mRNA | DQ446876.1 | - | 1 - 2 | 136 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |