Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118993.4 | NP_194580.1 | 2 - 3 | 307 | 93 | |
GenBank mRNA | AY093228.1 | - | 2 - 3 | 234 | 93 | |
BT000211.1 | - | 1 - 2 | 172 | 93 | ||
U66591.1 | - | 2 - 3 | 242 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |