Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118996.5 | NP_567812.1 | 11 - 12 | 1488 | 58 | |
GenBank mRNA | AY099715.1 | - | 11 - 12 | 1228 | 58 | |
AY943845.1 | - | 11 - 12 | 939 | 58 | ||
BT000286.1 | - | 11 - 12 | 939 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |