Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122014.4 | NP_197507.1 | 3 - 4 | 889 | 64 | |
GenBank mRNA | AK226922.1 | - | 3 - 4 | 853 | 64 | |
AY049280.1 | - | 3 - 4 | 870 | 64 | ||
AY090287.1 | - | 3 - 4 | 837 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |